Skip to Content
Merck
All Photos(1)

Key Documents

646075

Sigma-Aldrich

p-Tolylmagnesium bromide solution

0.5 M in diethyl ether

Synonym(s):

4-Methylphenylmagnesium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4MgBr
CAS Number:
Molecular Weight:
195.34
Beilstein:
636491
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in diethyl ether

SMILES string

Cc1ccc([Mg]Br)cc1

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup - Solvent

Flash Point(C)

-40 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derek E Williams et al.
Journal of the American Chemical Society, 140(24), 7611-7622 (2018-05-29)
Stimuli-responsive materials are vital for addressing emerging demands in the advanced technology sector as well as current industrial challenges. Here, we report for the first time that coordinative integration of photoresponsive building blocks possessing photochromic spiropyran and diarylethene moieties within
Ekaterina A Dolgopolova et al.
Journal of the American Chemical Society, 141(13), 5350-5358 (2019-03-07)
Electronic structure modulation of metal-organic frameworks (MOFs) through the connection of linker "wires" as a function of an external stimulus is reported for the first time. The established correlation between MOF electronic properties and photoisomerization kinetics as well as changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service