Skip to Content
Merck
All Photos(3)

Key Documents

374776

Sigma-Aldrich

4-Methoxybiphenyl

97%

Synonym(s):

4-Phenylanisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4OCH3
CAS Number:
Molecular Weight:
184.23
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

97%

form

powder, crystals or chunks

mp

86-90 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

COc1ccc(cc1)-c2ccccc2

InChI

1S/C13H12O/c1-14-13-9-7-12(8-10-13)11-5-3-2-4-6-11/h2-10H,1H3

InChI key

RHDYQUZYHZWTCI-UHFFFAOYSA-N

Application

4-Methoxybiphenyl has been used as a standard reagent whose fluorescence intensity is associated with the fluorescence characteristics of the products of derivatization reaction for aryl halides with phenylboronic acid (PBA).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Fry
Xenobiotica; the fate of foreign compounds in biological systems, 17(6), 751-758 (1987-06-01)
1. The metabolism of 4-methoxybiphenyl to 4-hydroxybiphenyl and its sulphate and glucuronic acid conjugates has been studied in rat isolated hepatocytes at various concentrations of 4-methoxybiphenyl. 2. The proportions of metabolites produced remained constant at concentrations of 4-methoxybiphenyl less than
Influence of the sulphation inhibitor, 2,6-dichloro-4-nitrophenol, on the production and conjugation, of 4-hydroxybiphenyl generated from 4-methoxybiphenyl by rat isolated hepatocytes.
J R Fry et al.
Biochemical pharmacology, 36(18), 3090-3092 (1987-09-15)
P Paterson et al.
Xenobiotica; the fate of foreign compounds in biological systems, 15(6), 493-502 (1985-06-01)
The rate of production of 4-hydroxybiphenyl from 4-methoxybiphenyl in hepatocytes isolated from untreated rats was essentially identical to that from biphenyl in hepatocytes isolated from rats pretreated with beta-naphthoflavone at 40 mg/kg. Similar results were obtained using liver microsomes isolated
A comparison of biphenyl 4-hydroxylation and 4-methoxybiphenyl O-demethylation in rat liver microsomes.
J R Fry
Biochemical pharmacology, 30(14), 1915-1919 (1981-07-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service