Merck
Antireflective disordered subwavelength structure on GaAs using spin-coated Ag ink mask.
Optics express (2012-10-06)
Chan Il Yeo, Ji Hye Kwon, Sung Jun Jang, Yong Tak Lee
RÉSUMÉ
We present a simple, cost-effective, large scale fabrication technique for antireflective disordered subwavelength structures (d-SWSs) on GaAs substrate by Ag etch masks formed using spin-coated Ag ink and subsequent inductively coupled plasma (ICP) etching process. The antireflection characteristics of GaAs d-SWSs rely on their geometric profiles, which were controlled by adjusting the distribution of Ag etch masks via changing the concentration of Ag atoms and the sintering temperature of Ag ink as well as the ICP etching conditions. The fabricated GaAs d-SWSs drastically reduced the reflection loss compared to that of bulk GaAs (>30%) in the wavelength range of 300-870 nm. The most desirable GaAs d-SWSs for practical solar cell applications exhibited a solar-weighted reflectance (SWR) of 2.12%, which is much lower than that of bulk GaAs (38.6%), and its incident angle-dependent SWR was also investigated.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Gallium arsenide, (single crystal substrate), <100>, diam. × thickness 2 in. × 0.5 mm