Merck
Sulfamethoxypyridazine-responsive pemphigoid nodularis: a report of two cases.
Journal of the American Academy of Dermatology (2005-07-16)
J E Gach, N J E Wilson, F Wojnarowska, A Ilchyshyn
RÉSUMÉ
Pemphigoid nodularis is a chronic and uncommon variant of bullous pemphigoid. The condition is characterized by the presence of prurigo nodularis-like lesions, possible history of blistering, and immunohistochemical findings of bullous pemphigoid. These patients are often unresponsive to conventional therapy with potent topical and systemic steroids and different immunosuppressive agents used alone or in combination. We describe two cases of pemphigoid nodularis treated successfully with sulfamethoxypyridazine without any adverse effects.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Sulfamethoxypyridazine, VETRANAL®, analytical standard