Key Documents
Z744090
Autoclave indicator tape
W × L 1.0 in. × 2,160 in.
Synonyme(s) :
Indicator tape
About This Item
Code UNSPSC :
41122410
Conditionnement
pack of 1 roll
Fabricant/nom de marque
Diversified Biotech ACTP-1001
Larg. × L
1.0 in. × 2,160 in.
Description générale
Printed diagonal stripes darken indicating a steam autoclave process is complete. Stripes darken after 5 minutes of exposure at 273°F (134°C) at steam pressure of 1.02 kg/cm2.