Principaux documents
Z719412
Sorenson low binding aerosol barrier tips
MultiGuard, maximum volume 20 μL
Synonyme(s) :
Aerosol Barrier Tips
About This Item
Code UNSPSC :
41121601
Nomenclature NACRES :
NB.01
Matériaux
colorless
polypropylene
Stérilité
sterile
Caractéristiques
DNase free
RNase free
barrier
Conditionnement
pack of 96 ea (Racked)
Fabricant/nom de marque
Sorenson 14210T
Volume maximum
20 μL
Type de liaison
low binding surface
Description générale
Tips feature a new surface technology designed for quantitative assays in which sample binding can significantly alter results or for critical small volume protocols that require a high degree of accuracy. Ideal for molecular biology applications, such as PCR. Inert hydrophobic barrier prevents pipettor contamination.
Informations légales
Sorenson is a trademark of Sorenson BioScience, Inc.