Principaux documents
Z659401
Bandelin SONOPULS ultrasonic homogenizer accessories
Indirect processing Cup horn BB 6 for use with HD 2200 homogenizer
Synonyme(s) :
sonication vessel
About This Item
Code UNSPSC :
41101520
Nomenclature NACRES :
NB.74
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bandelin 3605
Disponibilité
available only in EU
Description générale
BB6 for indirect irradiation with high energy. No contamination from cavitation erosion. No aerosoling with pathogenic or hazardous materials. Temperature regulation with circulating liquid coolants.