Principaux documents
Z659363
Bandelin SONOPULS ultrasonic homogenizer accessories
cooling vessel KG 5 for use with HD 2200 homogenizer
Synonyme(s) :
sonication vessel
About This Item
Code UNSPSC :
41101520
Nomenclature NACRES :
NB.74
Matériaux
glass
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bandelin 481
Disponibilité
available only in EU
Volume × D.I. × H
80 mL × 35 mm × 95 mm
Description générale
Cooling vessel KG for sonication of temperature-sensitive samples. The cooling jacket allows temperature control using liquid coolants during sonication.
Caractéristiques et avantages
  • Simultaneous sound and cooling
  • Optimal process observation