Key Documents
Z331678
BRAND® macro pipette controller
gray
Synonyme(s) :
brand pipette, macro pipette controller
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
gray
plastic (autoclavable)
silicone (autoclavable)
Caractéristiques
autoclavable
Fabricant/nom de marque
BRAND 26200
Volume
0.1-200 mL
Description générale
One single macro pipette controller covers the entire range of bulb and graduated pipettes from 0.1 to 200 mL. The conical silicone adapter offers a secure fit for the different diameters. The unit is fully autoclavable at 121°C (2 bar) according to DIN EN 285.
Supplied with a spare hydrophobic membrane filter which protects the system from liquid penetration.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Les clients ont également consulté
Slide 1 of 3
1 of 3