Principaux documents
TMO382019-0030
Nalgene® Certified Clean bottles
clear PETG, volume 30 mL, case of 72 ea
Synonyme(s) :
PETG bottle, certified nalgene bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
clear PETG
Conditionnement
case of 72 ea
Fabricant/nom de marque
Nalgene 382019-0030
Capacité
30 mL
Volume
30 mL
Description générale
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific Nalgene PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.
Caractéristiques et avantages
  • Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.
  • Exhibit excellent gas barrier properties and ensure pH stability for buffered solutions.
  • Resins are free of animal-derived components.
  • Sterile, tamper-resistant seal ensures product integrity prior to use.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries