Principaux documents
SYNB135415
Synthware vacuum desiccator
includes a ceramic plate and lid with stopcock/port, ID 150mm
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Kemtech B135415
Diamètre
150 mm
Description générale
Vacuum Desiccator, ID 150mm, Includes Lid & Ceramic Plate includes Accessories: A682440G
Application
kemetch; synthware; vacuum desiccator; desiccator
Caractéristiques et avantages
Organic Chemistry
Informations légales
Synthware is a trademark of Kemtech America Inc.