Principaux documents
MTCP4406
MTC Bio SureStand Multiple Pipette Stand
to hold, 6 x single pipettes OR 4 x multipipettes
Synonyme(s) :
pipette holder, pipette rack, plastic tube
About This Item
Code UNSPSC :
41122801
Nomenclature NACRES :
NB.02
Matériaux
acrylic
Caractéristiques
to hold 6 x single pipettes OR 4 x multipipettes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
MTC Bio P4406
Description générale
SureStand Pipette Stand for 6 pipettes, up to four multi-channels, acrylic
SureStand Pipette racks are unique in their ability to accommodate multi-channel pipettes without sacrificing any of the stability of a traditional acrylic rack. The patented “windows” allow the user to insert the multi-channel head longways through the rack while keeping the body of the pipette stable by securing the pipette at two points: under the finger hook and near the bottom of the handle. Each SureStand is also outfitted with four non-skid rubber “feet” for extra stability. SureStand is guaranteed to be the sturdiest and most versatile pipette rack in your lab.
Caractéristiques et avantages
  • Universal – Fits all popular pipettes
  • 5mm acrylic – 40% thicker
  • Non-skid rubber feet
  • Multi-channel “window” slots
  • US Patent No 872,300
Informations légales
MTC is a trademark of MTC Bio
SureStand is a trademark of MTC Bio