Principaux documents
MTCM4030
MTC Bio Rigid Inoculating Needles
pack of 1000 ea (50 x bags 20 ea), sterile
Synonyme(s) :
culture loop
About This Item
Code UNSPSC :
41161502
Nomenclature NACRES :
NB.15
Stérilité
sterile
Conditionnement
pack of 1000 ea (50 x bags 20 ea)
Fabricant/nom de marque
MTC Bio M4030
Description générale
Inoculating needles, rigid ABS, 50 sterile peel bags of 20 needles, 1000/pk
Caractéristiques et avantages
  • Sterile and disposable, no flaming
  • Uniform straking and inoculating
  • Hexagonal grip - improves control
  • Smooth loop surface - no gouging
Informations légales
MTC is a trademark of MTC Bio