Principaux documents
MTCA8815
MTC Bio Blu-Lite UHC Autoradiography Flim
W × L 8 in. × 10 in.
Synonyme(s) :
blotting film, western imaging film
About This Item
Code UNSPSC :
41105341
Nomenclature NACRES :
NB.15
Conditionnement
pack of 100 ea
Fabricant/nom de marque
MTC Bio A8815
Larg. × L
8 in. × 10 in.
Description générale
Blu-Lite UHC Autoradiography film, 8x10in, 100 sheets/box
Caractéristiques et avantages
  • Medium/dark blue base
  • Ideal for chemiluminescence
  • Ultra-high contrast
  • Ideal for 14C, 32P, 33P, 125I, and 35S
Informations légales
Blu-Lite is a trademark of MTC Bio
MTC is a trademark of MTC Bio