HS5360B
Benchtop counter
9 Key
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.22
Produits recommandés
L × l × H
12.5 × 3 × 2.2 in.
Catégories apparentées
Description générale
Mechanical benchtop counters are perfect for blood cells, bacterial colonies or other repetiitive counting.
Caractéristiques et avantages
- each key records up to 999
- total window keeps track of total number of strokes made on all other keys, bell rings when total reaches 100
- blood cell key labels