Key Documents
G3150
Sigma-Aldrich
Glass plates for vertical mini gel
8 cm × 10 cm
About This Item
Code UNSPSC :
41105312
Nomenclature NACRES :
NB.51
product name
Glass plates for vertical mini gel, 10 cm wide, Clear
Niveau de qualité
Taille
8 cm × 10 cm
Conditionnement
Set includes one rectangular plate with rounded bottom corners and one notched plate.
Compatibilité
for polyacrylamide gels