Principaux documents
EP5490047001
Rotor for Eppendorf® Concentrator Plus
F-35-6-30, to hold, 6 x 15 mL or 50 mL conical tubes
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.82
Caractéristiques
to hold 6 x 15 mL or 50 mL conical tubes
to hold 6 x 17 x 116 - 123 mm
to hold 6 x 29.5 x 116 - 123 mm
fixed angle rotor
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5490047001 (EU)
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models Concentrator Plus
Description générale
Selection of rotors for the Eppendorf Concentrator Plus.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG