Principaux documents
CLS352001
Corning® Falcon® Round Bottom Test Tubes
capacity 14 mL, cap, snap, sterile, individually wrapped, case of 500
Synonyme(s) :
Corning tube, cell culture tube
About This Item
Code UNSPSC :
41121700
Nomenclature NACRES :
NB.13
Matériaux
round polystyrene
Stérilité
sterile
Caractéristiques
cap snap
individually wrapped
Conditionnement
case of 500
Fabricant/nom de marque
Corning 352001
Capacité
14 mL
Application
Liquid Handling
Informations légales
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.