Principaux documents
BMSIPS2050PT2
Sigma-Aldrich
Benchmark Overhead Stirrer Propeller
centrifugal, PTFE-coated
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.72
Matériaux
PTFE-coated
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Benchmark Scientific IPS2050-P-T2
Diam. × L
85 mm × 350 mm , open
Description générale
Perfect for use is small mouth, round flasks, this PTFE coated propeller is designed for mid to high speed mixing.
Stiring rods and propellers to fit Benchmark range of OS-20L and OS-40L overhead stirrers.
Application
stirring, mixing