Key Documents
1.50083-M
Supelco
manu-CART Coupling unit for the coupling of two LiChroCART HPLC cartridges
About This Item
Code UNSPSC :
41115709
Nomenclature NACRES :
SB.52
Température de stockage
no temp limit
Niveau de qualité
Remarque sur l'analyse
Extent of testing: passes test
Informations légales
LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
MANU-CART is a trademark of Merck KGaA, Darmstadt, Germany