Z743307
Julabo® external Pt 100 bath sensors
m+r in-line sensor with m16x1 male connector, cable L 1.5 m
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
Produits recommandés
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Julabo 8981020
L du câble
1.5 m
Description générale
Pt 100 sensors enable you to reliably measure temperature within Julabo water baths.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.