Principaux documents
Z743302
Julabo® external Pt 100 bath sensors
L × diam. 200 mm × 6 mm, PTFE-coated stainless steel
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
Matériaux
PTFE-coated stainless steel
Description
without handle
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Julabo 8891032
L × diam.
200 mm × 6 mm
Description générale
Pt 100 sensors enable you to reliably measure temperature within Julabo water baths.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.