Principaux documents
Z742453
Mircrotube adapter for Orbi-Shaker MP and Incu-Mixer MP
holds 20 x 1.5-2 mL
About This Item
Code UNSPSC :
41103811
Nomenclature NACRES :
NB.73
Caractéristiques
holds 20 x 1.5-2 mL
Conditionnement
pkg of 1
Fabricant/nom de marque
Benchmark Scientific BT1500-A1520
Description générale
Microtube Adapter for microplate shakers BT1500 and H6004; shakers will fit max 4 adapters
Informations légales
Incu-Mixer is a trademark of Benchmark Scientific, Inc.
Orbi-Shaker is a trademark of Benchmark Scientific, Inc.