Key Documents
Z693812
LabJaws multi-rod connector
nickel plated
Synonyme(s) :
clamp holders, frame connectors
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.35
Matériaux
nickel plated
Fabricant/nom de marque
Troemner 916131
Capacité de la mâchoire
0-13 mm
Description générale
Maximum adjustment capability within one connector. Two adjustment rods fit into the oval hole in the connector, preventing turning when tightened. Side and front 13 mm holes allow creation of a variety of configurations.
Informations légales
LabJaws is a trademark of Henry Troemner LLC