Principaux documents
Z682780
Ace receiving flask,
capacity 50 mL, joint: ST/NS 35/20, glass
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
glass
round bottom flask
single-neck
Joint
joint: ST/NS35/20
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 1
Fabricant/nom de marque
Ace Glass 6902-234
Capacité
50 mL
Catégories apparentées
Description générale
The receiving flasks have the standard joint to fit the glassware used in rotary evaporators, and are available with poly-coating or plain glass.