Z682519
Ace vapor duct tube, for condenser "V" assembly
joint: ST/NS 29/42, L 45 mm, fits Büchi® models R-114/R-144
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Produits recommandés
Joint
joint: ST/NS29/42
Fabricant/nom de marque
Büchi equivalent, 32337
Longueur
45 mm
Catégories apparentées
Description générale
The vapor duct tube accessories are for Büchi condenser "V" assemblies, and will fit either Büchi Rotavapor model R-210/R-215 or R-114/R-144.
Combi-clips: not included.
Combi-clips: not included.
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG