Key Documents
Z618004
Binder drying oven accessories
Safety device (TWW) class 3.1
Synonyme(s) :
Binder 8012-0032
About This Item
Code UNSPSC :
41104512
Nomenclature NACRES :
NB.24
Caractéristiques
Temperature Safety Device for safe and consistent temperature regulation, Class 3.1 with visual alarm. For use with Binder units ED023 and ED400.
Conditionnement
pack of 1 ea
Disponibilité
not available in USA