Key Documents
Z556785
Sigma-Aldrich
Aldrich® fixed position Benchclamp
2-prong, size M
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.35
Taille
M
Capacité de la mâchoire
0-77 mm
Largeur de la fixation
23 mm
Description générale
Nickel-plated finish is corrosion resistant.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC