Key Documents
Z552593
Sigma-Aldrich
Aldrich® high-efficiency Liebig reflux condenser
®connectors, joint: ST/NS 24/40
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/40
Longueur
220 mm
Description générale
With SafetyBarb® pictured.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany