Key Documents
Z508470
Ace flat-bottom reaction flasks
capacity 4,000 mL, w/o O-ring groove
Synonyme(s) :
bottom flask, flat flask, reaction flasks
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
flat bottom
glass
single-neck
Stérilité
non-sterile
Caractéristiques
closure type Lid
Conditionnement
pack of 1
Fabricant/nom de marque
Ace Glass 6511-31
Capacité
4,000 mL
Description générale
Cylindrical heavy-wall reaction flask with a flat bottom rounded into side of wall, and extra-thick flat flange. No constriction at top facilitates introduction or removal of material and allows easy cleaning. Optional O-ring groove allows use of CAPFE O-rings rather than gaskets. Flask is supplied with one CAPFE O-ring. Order reaction flask heads and clamps separately.