Principaux documents
Z502529
Sigma-Aldrich
Glindemann PTFE sealing rings
(for 45/50 joints)
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.35
Joint
joint 45/50
Conditionnement
pkg of 50 ea
Fabricant/nom de marque
Glindemann K45
Plage de mesure de la température
≤250 °C
Description générale
  • This narrow ring seal is an alternative to a PTFE joint sleeve
  • Greaseless protection for air- and moisture- sensitive compounds
  • Can be used for prolonged storage under vacuum
  • Slide easily onto medium and full length taper joints
  • Can be used with Keck® clip and similar metal joint clips
Informations légales
Keck is a registered trademark of Adcuram Lab Technologies GmbH