Key Documents
Z415510
Sigma-Aldrich
Aldrich® Claisen distillation head with Vigreux indents
Joints: ST/NS 24/40 (3), H 380 mm, thermometer joint: ST/NS 10/30
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Joint
thermometer joint: ST/NS10/30
Joints: ST/NS24/40 (3)
Hauteur
380 mm
Profondeur d'immersion
75 mm
Description générale
Vigreux indents run the entire length of the column and act as the packing for the column.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC