Principaux documents
Z414344
Sigma-Aldrich
Aldrich® septum-inlet adapter with stopcock on side-arm
joint: ST/NS 14/20, PTFE stopcock
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Matériaux
PTFE stopcock
Joint
joint: ST/NS14/20
Description générale
Septum-Einlässe benötigen Septum Z10,072-2 oder Z12,435-4 sofern nicht mit * gekennzeichnet.
Liaison
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC