Key Documents
Z319368
Büchi® B-540/B-545 boiling point tubes
pack of 100 tubes
About This Item
Code UNSPSC :
41112203
Nomenclature NACRES :
NB.79
Conditionnement
pack of 100 tubes
Fabricant/nom de marque
Büchi 19007
Description générale
For an accurate determination of a melting point & boiling point.
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG