Principaux documents
Z302961
Quickfit® pear-shaped flasks
capacity 10 mL, joint: ST/NS 14/23
Synonyme(s) :
FP10/1
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
pear-shaped flask
single-neck
Joint
joint: ST/NS14/23
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Quickfit FP10/1
Disponibilité
not available in USA and Canada
Capacité
10 mL
Description générale
Single neck with ground-glass joint
Informations légales
Quickfit is a registered trademark of SciLabware Limited