Key Documents
Z275727
Nalgene® stirring rods
polypropylene
Synonyme(s) :
plastic stirring rod, stirring paddle
About This Item
Code UNSPSC :
41103800
Nomenclature NACRES :
NB.35
Matériaux
polypropylene
Conditionnement
pkg of 12 ea
Fabricant/nom de marque
Nalgene 6169-0010
Longueur
9 3/4 in.
Description générale
Combines the functions of a stirring rod and a rubber policeman. Paddles at each end, 5/8 in. W.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries