Principaux documents
Z231657
Duran® watch glasses
O.D. 250 mm
Synonyme(s) :
213216607
About This Item
Code UNSPSC :
41121801
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
DURAN®
D.E.
250 mm
Description générale
With fused edges. Manufactured from Duran borosilicate glass type 3.3 for maximum chemical and thermal shock resistance.
Caractéristiques et avantages
• Complies with DIN 12341• Manufactured from borosilicate glass 3.3• Reduced risk of injury due to fused rim• Heat resistant
Informations légales
DURAN is a registered trademark of DWK Life Sciences