Key Documents
Z220574
Sigma-Aldrich
NMR tube labels
temperature −40-150 °F, pkg of 25 ea
Synonyme(s) :
Labeling for NMR Tubes, Labels for NMR Tubes, NMR Label Set
About This Item
Code UNSPSC :
55121611
Nomenclature NACRES :
NB.35
Matériaux
vinyl
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Sigma-Aldrich
Paramètres
−40-150 °F temperature
Largeur
15 mm
Description générale
Self-laminating, vinyl labels with white, write-on surface resist oil, water, and solvent. Label wraps around 5mm tube for positive sample identification. Supplied with 26 labels on a card, 25 cards per pack.