Z147966
Aldrich® quick-distillation and sample collection tube
joint: ST/NS 35/45, thermometer joint: 10/18
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Produits recommandés
Joint
thermometer joint: 10/18
joint: ST/NS35/45
Catégories apparentées
Description générale
<ST/>35/45 joint, <ST/>10/18 thermometer joint. For distilling small samples. A 50mm immersion length thermometer is recommended.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC