Key Documents
Z136085
Bel-Bulb pipettor
vinyl bulb, fits 6-8 mm pipettes, L × diam. 70 mm × 5 mm
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
vinyl bulb
Caractéristiques
fits 6-8 mm pipettes
Conditionnement
pack of 2 ea
Fabricant/nom de marque
Bel-Art F37887-0000
L × diam.
70 mm × 5 mm
Description générale
Flexible vinyl bulb. A soft tapered chuck allows bulb to be held against pipette for suctioning.
Informations légales
Bel-Bulb is a trademark of SP Scienceware, a division of SP