Key Documents
Z104639
LDPE wash bottles
capacity 250 mL, Narrow-mouth
Synonyme(s) :
bottle lab, laboratory bottles, wash bottle
About This Item
Code UNSPSC :
41103209
Nomenclature NACRES :
NB.17
Matériaux
low-density polyethylene bottle
narrow-mouth bottle
natural bottle
polypropylene cap
Joint
Joints threaded neck (28)
Caractéristiques
label no label
vented
with cap
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Bel-Art F11618-0008
Capacité
250 mL
Description générale
Consist of one-piece cap and nozzle plus delivery tube. Deliver single drops or a needle-sharp stream. Tip may be cut to increase delivery amounts. Polyallomer cap.