Z102466
Luer-to-Luer connector
male Luer lock to male Luer lock, nickel plated
Synonyme(s) :
Luer connector, Luer fitting
About This Item
Produits recommandés
Matériaux
nickel plated
Fabricant/nom de marque
Popper 6177
Description générale
Water tested 45 psi.