Key Documents
Z102423
Double-ended needle
gauge 18, L 24 in.
About This Item
Code UNSPSC :
41122002
Nomenclature NACRES :
NB.03
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Cadence Science
Longueur
24 in.
Calibre
18
Description générale
304 SS. One noncoring tip, one flat-cut end.