Principaux documents
Y906603
Sigma-Aldrich
Valve
brass diaphragm, for CGA 110/170 right angle path, internal/external, right hand thread
About This Item
Code UNSPSC :
12142200
Nomenclature NACRES :
NA.12
Note: This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Matériaux
brass diaphragm
Niveau de qualité
Paramètres
3000 psi max. pressure
Longueur
3.31 in.
Poids
230 g
Raccords
for CGA 110/170 right angle path, internal/external, right hand thread
Catégories apparentées
Comparer avec des articles similaires
Voir la comparaison complète
Montrer les différences
1 of 4
Cet article
Y907227Y907219Y906468
Valve brass diaphragm, for CGA 110/170 right angle path, internal/external, right hand thread
Y906603
Valve
Valve brass diaphragm, for CGA 350 right angle path, external, left hand thread
Y907227
Valve
Valve brass diaphragm, for CGA 540 right angle path, internal, right hand thread
Y907219
Valve
Valve stainless steel diaphragm, for CGA 660 right angle path, external, right hand thread
Y906468
Valve
parameter
3000 psi max. pressure
parameter
2640 psi max. pressure
parameter
2640 psi max. pressure
parameter
2640 psi max. pressure
L
3.31 in.
L
4.62 in.
L
4.62 in.
L
4.62 in.
weight
230 g
weight
613 g
weight
694 g
weight
560 g
fitting
for CGA 110/170 right angle path, internal/external, right hand thread
fitting
for CGA 350 right angle path, external, left hand thread
fitting
for CGA 540 right angle path, internal, right hand thread
fitting
for CGA 660 right angle path, external, right hand thread
Autres remarques
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service