Principaux documents
TEM-LC400AU
Sigma-Aldrich
Lacey Carbon Supported Gold Grids
size 400 mesh, size 50 nm , hole size, box of 5 ×
Synonyme(s) :
Lacey Carbon Film, 400 Mesh, Gold, 5pk
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 5 ×
Taille
400 mesh
50 nm , hole size
Description générale
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.