Principaux documents
TEM-HC400-NI-100
Sigma-Aldrich
Holey Carbon Film Supported Nickel Grids
size 400 mesh, hole size 100 μm, box of 25
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 25
Taille de l'orifice
100 μm
Taille
400 mesh
Description générale
Holey Carbon Film Supported Nickel Grids, 400 Mesh, Average hole size is 100 micron, 25/box