Principaux documents
TEM-G200NI
Sigma-Aldrich
Glider Grids Standard Nickel Square Mesh
size 200 mesh, vial of 100
Synonyme(s) :
SQUARE 200 NESH NICKEL 100 VIAL
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
vial of 100
Taille
200 mesh
Description générale
Gilder Grids Standard Square Mesh: Nickel Grid, 200 mesh, 3.05mm diameter, 0.7 mil (18μm) thickness. Pitch 125μm, Hole 90μm, Bar 35μm