Principaux documents
SYNV210500
Synthware Dewar type vacuum trap
500 mL, stopcock: PTFE
Synonyme(s) :
Synthware V210500
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware V210500
Capacité
500 mL
Catégories apparentées
Description générale
Dry ice, Dewar style trap condenser designed for large volume solvent removal. Choice either glass stopcock for better vacuum or PTFE stopcock for grease-free operation. With 10 mm hose connections.
Informations légales
Synthware is a trademark of Kemtech America Inc.