Principaux documents
SYNS540004
Synthware high vacuum valve plug
size 0-4 mm, O-rings: Viton
Synonyme(s) :
Synthware S540004
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S540004
Description générale
Complete with control knob and PTFE stem having three O-rings.
Informations légales
Synthware is a trademark of Kemtech America Inc.