Principaux documents
SYNS321740
Synthware straight bore glass stopcock
bore size 4 mm, O.D. 10 mm stem, plug size 17/40
Synonyme(s) :
Synthware S321740, two way straigh bore stopcock
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S321740
Taille de l'alésage
4 mm
Description générale
Plugs and barrels are ground to be interchangeable, with 1:10 taper.
Informations légales
Synthware is a trademark of Kemtech America Inc.